Anti-IgG Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R & D Systems
F0110
220016-875EA 326.83 USD
220016-875
Anti-IgG Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR