Anti-IgG Donkey Polyclonal Antibody (Fluorescein)

Supplier: R & D Systems
F0109
220016-874EA 261.75 USD
220016-874
Anti-IgG Donkey Polyclonal Antibody (Fluorescein)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR