Anti-IgG Donkey Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R & D Systems
F0107
220016-872EA 326.83 USD
220016-872
Anti-IgG Donkey Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR