Anti-IgG Goat Polyclonal Antibody (Fluorescein)

Supplier: R & D Systems
F0103B
220016-869EA 229.94 USD
220016-869
Anti-IgG Goat Polyclonal Antibody (Fluorescein)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope: Heavy and Light Chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR