Anti-IgG Donkey Polyclonal Antibody

Supplier: R & D Systems
D-501-C-ABS
220014-793EA 163.42 USD
220014-793
Anti-IgG Donkey Polyclonal Antibody
Antibodies
Hidden

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR