Anti-IL18 Rat Monoclonal Antibody [clone: 93-10C]

Supplier: R & D Systems
D048-3
220014-775EA 626.18 USD
220014-775
Anti-IL18 Rat Monoclonal Antibody [clone: 93-10C]
Antibodies
Type: Primary
Antigen: IL18
Clonality: Monoclonal
Clone: 93-10C
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR