Anti-IL18 Rat Monoclonal Antibody [clone: 74]

Supplier: R & D Systems
D047-3
220014-774EA 631.97 USD
220014-774
Anti-IL18 Rat Monoclonal Antibody [clone: 74]
Antibodies
Type: Primary
Antigen: IL18
Clonality: Monoclonal
Clone: 74
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR