Anti-IL18 Rat Monoclonal Antibody [clone: 39-3F]

Supplier: R & D Systems
D046-3
220014-773EA 626.18 USD
220014-773
Anti-IL18 Rat Monoclonal Antibody [clone: 39-3F]
Antibodies
Type: Primary
Antigen: IL18
Clonality: Monoclonal
Clone: 39-3F
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR