Anti-IL18 Rat Monoclonal Antibody [clone: 159-12B]

Supplier: R & D Systems
D045-3
220014-771EA 637.75 USD
220014-771
Anti-IL18 Rat Monoclonal Antibody [clone: 159-12B]
Antibodies
Type: Primary
Antigen: IL18
Clonality: Monoclonal
Clone: 159-12B
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR