Anti-IL18 Mouse Monoclonal Antibody [clone: 125-2H]

Supplier: R & D Systems
D044-3
220014-770EA 631.97 USD
220014-770
Anti-IL18 Mouse Monoclonal Antibody [clone: 125-2H]
Antibodies
Type: Primary
Antigen: IL18
Clonality: Monoclonal
Clone: 125-2H
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR