Anti-IL18 Mouse Monoclonal Antibody [clone: 25-2G]

Supplier: R & D Systems
D043-3
220014-769EA 623.29 USD
220014-769
Anti-IL18 Mouse Monoclonal Antibody [clone: 25-2G]
Antibodies
Type: Primary
Antigen: IL18
Clonality: Monoclonal
Clone: 25-2G
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR