Anti-ICAM1 Mouse Monoclonal Antibody (Biotin) [clone: BBIG-I1 (11C81)]

Supplier: R & D Systems
BBA9
220014-705EA 633.42 USD
220014-705
Anti-ICAM1 Mouse Monoclonal Antibody (Biotin) [clone: BBIG-I1 (11C81)]
Antibodies
Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: BBIG-I1 (11C81)
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR