Anti-VCAM1 Mouse Monoclonal Antibody [clone: BBIG-V1]

Supplier: R & D Systems
BBA5
220014-701EA 588.58 USD
220014-701
Anti-VCAM1 Mouse Monoclonal Antibody [clone: BBIG-V1]
Antibodies
Adhesion Blockade

Type: Primary
Antigen: VCAM1
Clonality: Monoclonal
Clone: BBIG-V1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR