Anti-ITGA4 Mouse Monoclonal Antibody [clone: 2B4]

Supplier: R & D Systems
BBA37
220014-699EA 688.37 USD
220014-699
Anti-ITGA4 Mouse Monoclonal Antibody [clone: 2B4]
Antibodies
Adhesion Blockade

Type: Primary
Antigen: ITGA4
Clonality: Monoclonal
Clone: 2B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR