Anti-SELP Mouse Monoclonal Antibody (Fluorescein) [clone: 90]

Supplier: R & D Systems
BBA34
220014-697EA 349.97 USD
220014-697
Anti-SELP Mouse Monoclonal Antibody (Fluorescein) [clone: 90]
Antibodies
Type: Primary
Antigen: SELP
Clonality: Monoclonal
Clone: 90
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR