Anti-SELP Sheep Polyclonal Antibody

Supplier: R & D Systems
BBA32
220014-695EA 908.18 USD
220014-695
Anti-SELP Sheep Polyclonal Antibody
Antibodies
Adhesion Blockade

Type: Primary
Antigen: SELP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR