Anti-ICAM1 Mouse Monoclonal Antibody [clone: BBIG-I1 (11C81)]

Supplier: R & D Systems
BBA3
220014-693EA 623.29 USD
220014-693
Anti-ICAM1 Mouse Monoclonal Antibody [clone: BBIG-I1 (11C81)]
Antibodies
Adhesion Blockade

Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: BBIG-I1 (11C81)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR