Anti-ICAM3 Mouse Monoclonal Antibody [clone: Cal 3.10]

Supplier: R & D Systems
BBA29-200
102879-160EA 535.08 USD
102879-160
Anti-ICAM3 Mouse Monoclonal Antibody [clone: Cal 3.10]
Antibodies
Adhesion Blockade

Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: Cal 3.10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR