Anti-ICAM3 Mouse Monoclonal Antibody [clone: Cal 3.34]

Supplier: R & D Systems
BBA28
220014-691EA 633.42 USD
220014-691
Anti-ICAM3 Mouse Monoclonal Antibody [clone: Cal 3.34]
Antibodies
Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: Cal 3.34
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR