Anti-SELL Mouse Monoclonal Antibody [clone: 4G8]

Supplier: R & D Systems
BBA24
220014-688EA 642.09 USD
220014-688
Anti-SELL Mouse Monoclonal Antibody [clone: 4G8]
Antibodies
Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: 4G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR