Anti-VCAM1 Mouse Monoclonal Antibody (Fluorescein) [clone: BBIG-V3 (IE10)]

Supplier: R & D Systems
BBA22
220014-687EA 389.02 USD
220014-687
Anti-VCAM1 Mouse Monoclonal Antibody (Fluorescein) [clone: BBIG-V3 (IE10)]
Antibodies
Type: Primary
Antigen: VCAM1
Clonality: Monoclonal
Clone: BBIG-V3 (IE10)
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR