Anti-SELE Mouse Monoclonal Antibody (Fluorescein) [clone: BBIG-E5]

Supplier: R & D Systems
BBA21
220014-686EA 326.83 USD
220014-686
Anti-SELE Mouse Monoclonal Antibody (Fluorescein) [clone: BBIG-E5]
Antibodies
Type: Primary
Antigen: SELE
Clonality: Monoclonal
Clone: BBIG-E5
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR