Anti-ICAM1 Mouse Monoclonal Antibody (Fluorescein) [clone: BBIG-I1 (11C81)]

Supplier: R & D Systems
BBA20
220014-685EA 326.83 USD
220014-685
Anti-ICAM1 Mouse Monoclonal Antibody (Fluorescein) [clone: BBIG-I1 (11C81)]
Antibodies
Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: BBIG-I1 (11C81)
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR