Anti-SELE Mouse Monoclonal Antibody [clone: BBIG-E4]

Supplier: R & D Systems
BBA16
220014-680EA 540.86 USD
220014-680
Anti-SELE Mouse Monoclonal Antibody [clone: BBIG-E4]
Antibodies
Type: Primary
Antigen: SELE
Clonality: Monoclonal
Clone: BBIG-E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR