Anti-ICAM3 Mouse Monoclonal Antibody [clone: ICAM-3.3]

Supplier: R & D Systems
BBA15
220014-679EA 608.83 USD
220014-679
Anti-ICAM3 Mouse Monoclonal Antibody [clone: ICAM-3.3]
Antibodies
Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: ICAM-3.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR