Anti-CD44 (v3) Mouse Monoclonal Antibody [clone: 3G5]

Supplier: R & D Systems
BBA11
220014-677EA 665.23 USD
220014-677
Anti-CD44 (v3) Mouse Monoclonal Antibody [clone: 3G5]
Antibodies
Type: Primary
Antigen: CD44 (v3)
Clonality: Monoclonal
Clone: 3G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR