Anti-CD62E/CD62P Mouse Monoclonal Antibody [clone: BBIG-E6 (13D5)]

Supplier: R & D Systems
BBA1
220014-675EA 584.25 USD
220014-675
Anti-CD62E/CD62P Mouse Monoclonal Antibody [clone: BBIG-E6 (13D5)]
Antibodies
Adhesion Blockade

Type: Primary
Antigen: CD62E/CD62P
Clonality: Monoclonal
Clone: BBIG-E6 (13D5)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR