Anti-GMCSF1 Mouse Monoclonal Antibody (Biotin) [clone: 159319]

Supplier: R & D Systems
BAM9873
220014-673EA 728.86 USD
220014-673
Anti-GMCSF1 Mouse Monoclonal Antibody (Biotin) [clone: 159319]
Antibodies
Type: Primary
Antigen Symbol: GM-CSF
Antigen Name:
Antigen Synonyms:
Clonality: Monoclonal
Clone: 159319
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR