Anti-TIMP3 Mouse Monoclonal Antibody (Biotin) [clone: 277128]

Supplier: R & D Systems
BAM9731
220014-672EA 746.22 USD
220014-672
Anti-TIMP3 Mouse Monoclonal Antibody (Biotin) [clone: 277128]
Antibodies
Type: Primary
Antigen: TIMP3
Clonality: Monoclonal
Clone: 277128
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR