Anti-EPO Rat Monoclonal Antibody (Biotin) [clone: 148409]

Supplier: R & D Systems
BAM9592
220014-671EA 746.22 USD
220014-671
Anti-EPO Rat Monoclonal Antibody (Biotin) [clone: 148409]
Antibodies
Type: Primary
Antigen: EPO
Clonality: Monoclonal
Clone: 148409
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR