Anti-CDH5 Mouse Monoclonal Antibody (Biotin) [clone: 123413]

Supplier: R & D Systems
BAM9381
220014-670EA 594.37 USD
220014-670
Anti-CDH5 Mouse Monoclonal Antibody (Biotin) [clone: 123413]
Antibodies
Type: Primary
Antigen: CDH5
Clonality: Monoclonal
Clone: 123413
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR