Anti-ADAM17 Mouse Monoclonal Antibody (Biotin) [clone: 224964]

Supplier: R & D Systems
BAM9303
220014-669EA 714.4 USD
220014-669
Anti-ADAM17 Mouse Monoclonal Antibody (Biotin) [clone: 224964]
Antibodies
Type: Primary
Antigen: ADAM17
Clonality: Monoclonal
Clone: 224964
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR