Anti-ACE Mouse Monoclonal Antibody (Biotin) [clone: 171417]

Supplier: R & D Systems
BAM929
220014-668EA 543.75 USD
220014-668
Anti-ACE Mouse Monoclonal Antibody (Biotin) [clone: 171417]
Antibodies
Type: Primary
Antigen: ACE
Clonality: Monoclonal
Clone: 171417
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR