Anti-EDA Mouse Monoclonal Antibody (Biotin) [clone: 174817]

Supplier: R & D Systems
BAM9221
220014-667EA 681.14 USD
220014-667
Anti-EDA Mouse Monoclonal Antibody (Biotin) [clone: 174817]
Antibodies
Type: Primary
Antigen: EDA
Clonality: Monoclonal
Clone: 174817
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR