Anti-IL12 + IL23 Mouse Monoclonal Antibody (Biotin) [clone: 116211]

Supplier: R & D Systems
BAM9122
220014-666EA 714.4 USD
220014-666
Anti-IL12 + IL23 Mouse Monoclonal Antibody (Biotin) [clone: 116211]
Antibodies
Type: Primary
Antigen: IL12 + IL23
Clonality: Monoclonal
Clone: 116211
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR