Anti-MMP9 Rat Monoclonal Antibody (Biotin) [clone: 116125]

Supplier: R & D Systems
BAM909
220014-665EA 746.22 USD
220014-665
Anti-MMP9 Rat Monoclonal Antibody (Biotin) [clone: 116125]
Antibodies
Type: Primary
Antigen: MMP9
Clonality: Monoclonal
Clone: 116125
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR