Anti-CXCL14 Mouse Monoclonal Antibody (Biotin) [clone: 131107]

Supplier: R & D Systems
BAM8661
220014-663EA 728.86 USD
220014-663
Anti-CXCL14 Mouse Monoclonal Antibody (Biotin) [clone: 131107]
Antibodies
Type: Primary
Antigen: CXCL14
Clonality: Monoclonal
Clone: 131107
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR