Anti-IL22 Mouse Monoclonal Antibody (Biotin) [clone: 142928]

Supplier: R & D Systems
BAM7821
220014-661EA 740.43 USD
220014-661
Anti-IL22 Mouse Monoclonal Antibody (Biotin) [clone: 142928]
Antibodies
Type: Primary
Antigen: IL22
Clonality: Monoclonal
Clone: 142928
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR