Anti-IFNG Mouse Monoclonal Antibody (Biotin) [clone: 142508]

Supplier: R & D Systems
BAM7811
102879-152EA 746.22 USD
102879-152
Anti-IFNG Mouse Monoclonal Antibody (Biotin) [clone: 142508]
Antibodies
Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: 142508
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR