Anti-FLT3 Rat Monoclonal Antibody (Biotin) [clone: 113308]

Supplier: R & D Systems
BAM7681
220014-660EA 543.75 USD
220014-660
Anti-FLT3 Rat Monoclonal Antibody (Biotin) [clone: 113308]
Antibodies
Type: Primary
Antigen Symbol: FLT3
Antigen Name: FMS-like tyrosine kinase 3
Antigen Synonyms: B230315G04, CD135, Flk-2, Flk2, Flt-3, Ly72, wmfl
Clonality: Monoclonal
Clone: 113308
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR