Anti-TYRO3 Rat Monoclonal Antibody (Biotin) [clone: 109646]

Supplier: R & D Systems
BAM759
220014-658EA 714.4 USD
220014-658
Anti-TYRO3 Rat Monoclonal Antibody (Biotin) [clone: 109646]
Antibodies
Type: Primary
Antigen: TYRO3
Clonality: Monoclonal
Clone: 109646
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR