Anti-BMP4 Mouse Monoclonal Antibody (Biotin) [clone: 66108]

Supplier: R & D Systems
BAM7572
220014-657EA 746.22 USD
220014-657
Anti-BMP4 Mouse Monoclonal Antibody (Biotin) [clone: 66108]
Antibodies
Type: Primary
Antigen: BMP4
Clonality: Monoclonal
Clone: 66108
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR