Anti-EDAR Rat Monoclonal Antibody (Biotin) [clone: 132144]

Supplier: R & D Systems
BAM7452
220014-655EA 728.86 USD
220014-655
Anti-EDAR Rat Monoclonal Antibody (Biotin) [clone: 132144]
Antibodies
Type: Primary
Antigen: EDAR
Clonality: Monoclonal
Clone: 132144
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR