Anti-CD6 Rat Monoclonal Antibody (Biotin) [clone: 96123]

Supplier: R & D Systems
BAM727
220014-654EA 728.86 USD
220014-654
Anti-CD6 Rat Monoclonal Antibody (Biotin) [clone: 96123]
Antibodies
Type: Primary
Antigen: CD6
Clonality: Monoclonal
Clone: 96123
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR