Anti-TNFSF18 Mouse Monoclonal Antibody (Biotin) [clone: 109117]

Supplier: R & D Systems
BAM6943
220014-649EA 762.12 USD
220014-649
Anti-TNFSF18 Mouse Monoclonal Antibody (Biotin) [clone: 109117]
Antibodies
Type: Primary
Antigen: TNFSF18
Clonality: Monoclonal
Clone: 109117
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR