Anti-BDNF Mouse Monoclonal Antibody (Biotin) [clone: 37141]

Supplier: R & D Systems
BAM648
220014-645EA 753.45 USD
220014-645
Anti-BDNF Mouse Monoclonal Antibody (Biotin) [clone: 37141]
Antibodies
Type: Primary
Antigen: BDNF
Clonality: Monoclonal
Clone: 37141
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR