Anti-VCAM1 Rat Monoclonal Antibody (Biotin) [clone: 112731]

Supplier: R & D Systems
BAM6433
220014-644EA 723.08 USD
220014-644
Anti-VCAM1 Rat Monoclonal Antibody (Biotin) [clone: 112731]
Antibodies
Type: Primary
Antigen: VCAM1
Clonality: Monoclonal
Clone: 112731
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR