Anti-TGF-beta 3 Mouse Monoclonal Antibody (Biotin) [clone: 44922]

Supplier: R & D Systems
BAM643
220014-643EA 762.12 USD
220014-643
Anti-TGF-beta 3 Mouse Monoclonal Antibody (Biotin) [clone: 44922]
Antibodies
Type: Primary
Antigen Symbol: TGF-beta 3
Antigen Name:
Antigen Synonyms:
Clonality: Monoclonal
Clone: 44922
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR