Anti-alpha 2u-Globulin Mouse Monoclonal Antibody [clone: 129736] (Biotin)

Supplier: R & D Systems
BAM586
220014-641EA 728.86 USD
220014-641
Anti-alpha 2u-Globulin Mouse Monoclonal Antibody [clone: 129736] (Biotin)
Antibodies
Type: Primary
Antigen Symbol:
Antigen Name: alpha 2u-Globulin
Antigen Synonyms:
Clonality: Monoclonal
Clone: 129736
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR