Anti-ACVR1C Mouse Monoclonal Antibody (Biotin) [clone: 118311]

Supplier: R & D Systems
BAM5771
220014-640EA 543.75 USD
220014-640
Anti-ACVR1C Mouse Monoclonal Antibody (Biotin) [clone: 118311]
Antibodies
Type: Primary
Antigen: ACVR1C
Clonality: Monoclonal
Clone: 118311
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR